Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kif11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016116.3 | NP_001016116.2 | 6 - 7 | 746 | 67 | |
XM_012966561.1 | XP_012822015.1 | 7 - 8 | 906 | 67 | ||
GenBank mRNA | BC166147.1 | - | 6 - 7 | 677 | 67 | |
CR762094.2 | - | 6 - 7 | 746 | 67 | ||
EF524557.1 | - | 5 - 6 | 565 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |