Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016126.2 | NP_001016126.1 | 7 - 8 | 823 | 91 | |
GenBank mRNA | BC098087.1 | - | 7 - 8 | 798 | 91 | |
BC158989.1 | - | 7 - 8 | 801 | 91 | ||
BC170718.1 | - | 7 - 8 | 823 | 91 | ||
BC170720.1 | - | 7 - 8 | 823 | 91 | ||
CR762063.2 | - | 7 - 8 | 823 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |