Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016174.2 | NP_001016174.1 | 2 - 3 | 448 | 73 | |
GenBank mRNA | CR761871.2 | - | 2 - 3 | 448 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |