Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016177.2 | NP_001016177.1 | 1 - 2 | 226 | 88 | |
GenBank mRNA | BC135884.1 | - | 1 - 2 | 226 | 88 | |
CR761863.2 | - | 1 - 2 | 226 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |