Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016177.2 | NP_001016177.1 | 2 - 3 | 370 | 98 | |
XM_012968372.2 | XP_012823826.1 | 3 - 4 | 98 | |||
GenBank mRNA | BC135884.1 | - | 2 - 3 | 98 | ||
CR761863.2 | - | 2 - 3 | 370 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |