Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016218.2 | NP_001016218.1 | 485 | 71 | ||
GenBank mRNA | BC171291.1 | - | 485 | 71 | ||
BC171293.1 | - | 485 | 71 | |||
CR926314.2 | - | 485 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis |