Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccnh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016256.2 | NP_001016256.1 | 6 - 7 | 759 | 85 | |
GenBank mRNA | BC118698.1 | - | 6 - 7 | 789 | 85 | |
CR761723.2 | - | 6 - 7 | 759 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |