Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dctd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016265.2 | NP_001016265.1 | 3 - 4 | 294 | 91 | |
GenBank mRNA | CR761692.2 | - | 3 - 4 | 294 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |