Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dcun1d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016303.2 | NP_001016303.1 | 7 - 8 | 607 | 79 | |
XM_012956942.2 | XP_012812396.1 | 7 - 8 | 641 | 79 | ||
XM_018091244.1 | XP_017946733.1 | 7 - 8 | 1620 | 79 | ||
GenBank mRNA | BC167304.1 | - | 7 - 8 | 641 | 79 | |
BC171061.1 | - | 6 - 7 | 594 | 79 | ||
CR761510.2 | - | 7 - 8 | 607 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |