Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anapc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016327.2 | NP_001016327.1 | 3 - 4 | 458 | 105 | |
GenBank mRNA | BC136241.1 | - | 3 - 4 | 466 | 105 | |
BC155384.1 | - | 3 - 4 | 434 | 105 | ||
CR761429.2 | - | 3 - 4 | 458 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |