Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pabpn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016352.2 | NP_001016352.1 | 6 - 7 | 678 | 92 | |
XM_012961085.2 | XP_012816539.1 | 7 - 8 | 997 | 92 | ||
XM_012961086.2 | XP_012816540.1 | 7 - 8 | 764 | 92 | ||
XM_012961087.2 | XP_012816541.1 | 7 - 8 | 783 | 92 | ||
GenBank mRNA | BC121546.1 | - | 7 - 8 | 889 | 92 | |
CR761333.2 | - | 6 - 7 | 678 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |