Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016389.2 | NP_001016389.1 | 8 - 9 | 785 | 99 | |
XM_012968399.2 | XP_012823853.1 | 8 - 9 | 868 | 99 | ||
XM_012968400.2 | XP_012823854.1 | 8 - 9 | 851 | 99 | ||
XM_012968401.2 | XP_012823855.1 | 8 - 9 | 861 | 99 | ||
XM_012968402.2 | XP_012823856.1 | 8 - 9 | 868 | 99 | ||
XM_012968403.2 | XP_012823857.1 | 8 - 9 | 865 | 99 | ||
XM_012968404.2 | XP_012823858.1 | 8 - 9 | 861 | 99 | ||
XM_012968405.2 | XP_012823859.1 | 8 - 9 | 868 | 99 | ||
XM_018096253.1 | XP_017951742.1 | 8 - 9 | 826 | 99 | ||
GenBank mRNA | BC090121.1 | - | 7 - 8 | 769 | 99 | |
BC157299.1 | - | 7 - 8 | 772 | 99 | ||
BC158945.1 | - | 8 - 9 | 866 | 99 | ||
CR761189.2 | - | 8 - 9 | 785 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |