Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ca12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016431.2 | NP_001016431.1 | 5 - 6 | 622 | 86 | |
GenBank mRNA | BC135174.2 | - | 5 - 6 | 555 | 86 | |
CR761051.2 | - | 5 - 6 | 622 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |