Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mreg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016432.2 | NP_001016432.1 | 4 - 5 | 416 | 92 | |
XM_012970420.1 | XP_012825874.1 | 14 - 15 | 1801 | 92 | ||
XM_012970423.2 | XP_012825877.1 | 5 - 6 | 591 | 92 | ||
XM_018097106.1 | XP_017952595.1 | 4 - 5 | 781 | 92 | ||
GenBank mRNA | BC122004.1 | - | 4 - 5 | 349 | 92 | |
CR761048.2 | - | 4 - 5 | 416 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |