Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aldh3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016537.2 | NP_001016537.1 | 4 - 5 | 727 | 110 | |
XM_012956976.2 | XP_012812430.1 | 5 - 6 | 857 | 110 | ||
XM_012956977.2 | XP_012812431.1 | 5 - 6 | 825 | 110 | ||
XM_012956978.2 | XP_012812432.1 | 5 - 6 | 856 | 110 | ||
XM_012956979.2 | XP_012812433.1 | 5 - 6 | 836 | 110 | ||
XM_012956980.2 | XP_012812434.1 | 5 - 6 | 807 | 110 | ||
GenBank mRNA | BC091032.1 | - | 4 - 5 | 727 | 110 | |
CR760779.2 | - | 4 - 5 | 727 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |