Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aurka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016570.2 | NP_001016570.1 | 803 | 67 | ||
GenBank mRNA | BC169141.1 | - | 852 | 67 | ||
CR760668.2 | - | 803 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis |