Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dusp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016576.2 | NP_001016576.1 | 5 - 6 | 804 | 83 | |
GenBank mRNA | BC160459.1 | - | 6 - 7 | 857 | 83 | |
CR760642.2 | - | 5 - 6 | 804 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |