Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016584.2 | NP_001016584.1 | 6 - 7 | 410 | 73 | |
XM_012966612.2 | XP_012822066.1 | 6 - 7 | 578 | 73 | ||
GenBank mRNA | CR760621.2 | - | 6 - 7 | 410 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |