Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pole4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016605.2 | NP_001016605.1 | 2 - 3 | 312 | 80 | |
GenBank mRNA | BC161599.1 | - | 2 - 3 | 316 | 80 | |
BC170722.1 | - | 2 - 3 | 311 | 80 | ||
CR760539.2 | - | 2 - 3 | 312 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |