Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnali1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016687.2 | NP_001016687.1 | 1 - 2 | 115 | 82 | |
GenBank mRNA | BC135942.1 | - | 1 - 2 | 114 | 82 | |
CR760194.2 | - | 1 - 2 | 115 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |