Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdk2ap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016730.2 | NP_001016730.1 | 1 - 2 | 74 | 74 | |
GenBank mRNA | BC084911.1 | - | 1 - 2 | 177 | 74 | |
BC121523.1 | - | 1 - 2 | 93 | 74 | ||
BC167332.1 | - | 1 - 2 | 285 | 74 | ||
CR760030.2 | - | 1 - 2 | 74 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |