Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: letm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016801.2 | NP_001016801.1 | 6 - 7 | 815 | 108 | |
XM_012959045.2 | XP_012814499.1 | 5 - 6 | 984 | 108 | ||
GenBank mRNA | CR855606.2 | - | 6 - 7 | 815 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Xenopus tropicalis Probe spans exons |