Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: herpud2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016818.2 | NP_001016818.1 | 1 - 2 | 386 | 107 | |
XM_012964732.2 | XP_012820186.1 | 2 - 3 | 650 | 107 | ||
XM_012964733.1 | XP_012820187.1 | 2 - 3 | 513 | 107 | ||
XM_012964734.2 | XP_012820188.1 | 2 - 3 | 650 | 107 | ||
GenBank mRNA | CR855549.2 | - | 1 - 2 | 386 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Xenopus tropicalis Probe spans exons |