Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ciao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016887.2 | NP_001016887.1 | 7 - 8 | 945 | 65 | |
XM_012959054.2 | XP_012814508.1 | 7 - 8 | 943 | 65 | ||
XM_012959056.2 | XP_012814510.1 | 7 - 8 | 875 | 65 | ||
XM_012959057.2 | XP_012814511.1 | 7 - 8 | 868 | 65 | ||
GenBank mRNA | BC127279.1 | - | 7 - 8 | 939 | 65 | |
CR848572.2 | - | 7 - 8 | 945 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |