Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gtf2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016908.2 | NP_001016908.1 | 6 - 7 | 652 | 96 | |
GenBank mRNA | CR848470.2 | - | 6 - 7 | 652 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Xenopus tropicalis Probe spans exons |