Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnajc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016942.2 | NP_001016942.1 | 2 - 3 | 167 | 64 | |
XM_012964760.2 | XP_012820214.1 | 2 - 3 | 117 | 64 | ||
GenBank mRNA | BC155976.1 | - | 2 - 3 | 167 | 64 | |
CR761895.2 | - | 2 - 3 | 167 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |