Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tdp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016944.2 | NP_001016944.1 | 4 - 5 | 592 | 89 | |
XM_012964761.1 | XP_012820215.1 | 4 - 5 | 602 | 89 | ||
GenBank mRNA | BC166146.1 | - | 4 - 5 | 594 | 89 | |
CR761829.2 | - | 4 - 5 | 592 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |