Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: znrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016977.2 | NP_001016977.1 | 4 - 5 | 674 | 82 | |
GenBank mRNA | CR848382.2 | - | 4 - 5 | 674 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Xenopus tropicalis Probe spans exons |