Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017097.2 | NP_001017097.1 | 4 - 5 | 760 | 66 | |
XM_012963303.2 | XP_012818757.1 | 4 - 5 | 700 | 66 | ||
XM_012963305.2 | XP_012818759.1 | 4 - 5 | 837 | 66 | ||
XM_018093690.1 | XP_017949179.1 | 5 - 6 | 1015 | 66 | ||
GenBank mRNA | BC157270.1 | - | 4 - 5 | 675 | 66 | |
CR761309.2 | - | 4 - 5 | 760 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |