Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: uchl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017120.2 | NP_001017120.1 | 5 - 6 | 739 | 70 | |
GenBank mRNA | BC135703.1 | - | 5 - 6 | 485 | 70 | |
CR761162.2 | - | 5 - 6 | 739 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |