Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elovl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017257.3 | NP_001017257.2 | 1 - 2 | 154 | 71 | |
GenBank mRNA | CR760034.2 | - | 1 - 2 | 222 | 71 | |
CU075326.1 | - | 1 - 2 | 154 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Xenopus tropicalis Probe spans exons |