Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: polr3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017346.2 | NP_001017346.1 | 5 - 6 | 530 | 97 | |
XM_012959121.2 | XP_012814575.1 | 5 - 6 | 573 | 97 | ||
GenBank mRNA | BC121511.1 | - | 5 - 6 | 485 | 97 | |
CR855444.2 | - | 5 - 6 | 530 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |