Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spsb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017350.2 | NP_001017350.1 | 3 - 4 | 522 | 94 | |
XM_012970482.2 | XP_012825936.1 | 4 - 5 | 597 | 94 | ||
XM_012970483.2 | XP_012825937.1 | 4 - 5 | 700 | 94 | ||
XM_012970484.2 | XP_012825938.1 | 4 - 5 | 594 | 94 | ||
XM_018097133.1 | XP_017952622.1 | 4 - 5 | 587 | 94 | ||
GenBank mRNA | BC160472.1 | - | 4 - 5 | 570 | 94 | |
CR848600.2 | - | 3 - 4 | 522 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |