Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035118.1 | NP_001030290.1 | 3 - 4 | 1226 | 99 | |
GenBank mRNA | BC170914.1 | - | 3 - 4 | 1178 | 99 | |
BC170916.1 | - | 3 - 4 | 1178 | 99 | ||
CR926197.2 | - | 3 - 4 | 1226 | 99 | ||
DQ124205.1 | - | 3 - 4 | 1226 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |