Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cct7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037270.1 | NP_001032347.1 | 7 - 8 | 820 | 88 | |
GenBank mRNA | BC068214.1 | - | 7 - 8 | 869 | 88 | |
BC089710.1 | - | 7 - 8 | 820 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |