Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203937.3 | NP_989268.3 | 5 - 6 | 909 | 98 | |
GenBank mRNA | BC064215.1 | - | 5 - 6 | 901 | 98 | |
CR760996.2 | - | 5 - 6 | 892 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |