Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yipf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015932.3 | NP_001015932.1 | 2 - 3 | 179 | 93 | |
GenBank mRNA | BC122940.1 | - | 2 - 3 | 179 | 93 | |
CR926375.2 | - | 2 - 3 | 175 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |