Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trappc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016642.3 | NP_001016642.1 | 4 - 5 | 382 | 111 | |
XM_012956993.2 | XP_012812447.1 | 4 - 5 | 547 | 111 | ||
GenBank mRNA | BC121995.1 | - | 4 - 5 | 382 | 111 | |
CR760400.2 | - | 3 - 4 | 372 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |