Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: znhit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017056.3 | NP_001017056.2 | 3 - 4 | 584 | 75 | |
GenBank mRNA | BC121538.1 | - | 3 - 4 | 584 | 75 | |
CR761732.2 | - | 3 - 4 | 290 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |