Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp6v0d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017310.3 | NP_001017310.1 | 6 - 7 | 821 | 87 | |
GenBank mRNA | BC125665.1 | - | 6 - 7 | 821 | 87 | |
CR855501.2 | - | 6 - 7 | 833 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |