Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043347.1 | NP_001036812.1 | 6 - 7 | 739 | 72 | |
GenBank mRNA | BC087820.1 | - | 6 - 7 | 675 | 72 | |
BC121840.1 | - | 6 - 7 | 739 | 72 | ||
CR942510.2 | - | 6 - 7 | 715 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |