Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vma21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044463.1 | NP_001037928.1 | 2 - 3 | 230 | 80 | |
GenBank mRNA | BC159038.1 | - | 2 - 3 | 236 | 80 | |
CR761257.2 | - | 2 - 3 | 230 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |