Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zc3h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044484.3 | NP_001037949.3 | 5 - 6 | 1888 | 103 | |
XM_018094683.1 | XP_017950172.1 | 5 - 6 | 2009 | 103 | ||
GenBank mRNA | AY887083.1 | - | 5 - 6 | 1777 | 103 | |
BC121894.1 | - | 5 - 6 | 1888 | 103 | ||
DN005168.1 | - | 2 - 3 | 213 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |