Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ociad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044487.1 | NP_001037952.1 | 5 - 6 | 403 | 77 | |
GenBank mRNA | BC076708.1 | - | 5 - 6 | 274 | 77 | |
CR760011.2 | - | 5 - 6 | 403 | 77 | ||
CR761639.2 | - | 5 - 6 | 384 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |