Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrps12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044523.1 | NP_001037988.1 | 2 - 3 | 95 | 84 | |
XM_018096287.1 | XP_017951776.1 | 2 - 3 | 81 | |||
GenBank mRNA | BC135465.1 | - | 1 - 2 | 78 | 84 | |
CR762089.3 | - | 2 - 3 | 95 | 84 | ||
CT025408.2 | - | 2 - 3 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |