Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tor1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045631.1 | NP_001039096.1 | 3 - 4 | 684 | 74 | |
GenBank mRNA | BC160421.1 | - | 3 - 4 | 688 | 74 | |
CR942373.2 | - | 3 - 4 | 684 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |