Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ipo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045658.1 | NP_001039123.1 | 29 - 30 | 2878 | 85 | |
GenBank mRNA | BC168434.1 | - | 29 - 30 | 2792 | 85 | |
CR942469.2 | - | 29 - 30 | 2878 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |