Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam83c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045662.1 | NP_001039127.1 | 2 - 3 | 889 | 92 | |
XM_012952528.1 | XP_012807982.1 | 2 - 3 | 889 | 92 | ||
GenBank mRNA | BC168600.1 | - | 2 - 3 | 823 | 92 | |
CR942480.1 | - | 2 - 3 | 889 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |