Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045689.2 | NP_001039154.1 | 8 - 9 | 957 | 103 | |
GenBank mRNA | CR942586.2 | - | 7 - 8 | 886 | 103 | |
DR897520.1 | - | 3 - 4 | 697 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Xenopus tropicalis Probe spans exons |