Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045722.1 | NP_001039187.1 | 2 - 3 | 509 | 81 | |
GenBank mRNA | BC160529.1 | - | 1 - 2 | 517 | 81 | |
CR855534.2 | - | 2 - 3 | 509 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |